2,5-二溴硝基苯 97%
供应商: Thermo Fisher Scientific
|
Warning
|
Synonyms:
2,5-Dibromonitrobenzene
112860250
112861000
ACRO112860250EA
0
CNY
ACRO112860250
ACRO112861000
2,5-二溴硝基苯 97%
2,5-二溴硝基苯
分子式:
C₆H₃Br₂NO₂ 溶点: 81…85 °C 密度: 2,374 g/cm³ |
MDL编号:
MFCD00007046 CAS号: 3460-18-2 EINECS: 222-404-8 UN号: 2811 |
产品技术规格
Appearance | Yellow Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 81 to 85 °C |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...