Nitrosium hexafluorophosphate 95%
供应商: Thermo Fisher Scientific
|
Danger
|
Synonyms:
Nitrosonium hexafluorophosphate, Nitrosyl hexafluorophosphate
383730050
383730250
ACRO383730050EA
0
CNY
ACRO383730050
ACRO383730250
Nitrosium hexafluorophosphate 95%
Nitrosium hexafluorophosphate
分子式:
F₆NOP 存储条件: Ambient |
MDL编号:
MFCD00040324 CAS号: 16921-91-8 EINECS: 240-980-9 UN号: 3260 ADR: 8,III |
产品技术规格
Appearance | Colorless to white Crystals |
Assay | ≥94 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...