肉桂酸苄酯 99%

供应商: Thermo Fisher Scientific

Synonyms: 肉桂酸苯甲酯

215291000 215295000 215290050
ACRO215291000EA 0 CNY
ACRO215291000 ACRO215295000 ACRO215290050
肉桂酸苄酯 99%
肉桂酸苄酯
分子式: C₁₆H₁₄O₂
分子量: 238.29 g/mol
沸点: 195…200 °C (5 mmHg)
溶点: 33…37 °C
闪点: >112 °C
MDL编号: MFCD00004789
CAS号: 103-41-3
EINECS: 203-109-3

现在订购

产品技术规格

Appearance Clear colorless to yellow Crystalline mass or liquid after melting
Infrared spectrum Conforms
Melting point 33 to 37 °C
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR