氧化二乙酰丙酮合钒 99%
供应商: Thermo Fisher Scientific
|
Warning
|
Synonyms:
VO(acac)2, 氧化钒(IV)乙酰丙酮
199590500
199592500
ACRO199590500EA
0
CNY
ACRO199590500
ACRO199592500
氧化二乙酰丙酮合钒 99%
氧化二乙酰丙酮合钒
分子式:
C₁₀H₁₄O₅V 分子量: 265.16 g/mol 溶点: 256…259 °C 闪点: 79 °C 存储条件: Ambient |
MDL编号:
MFCD00000032 CAS号: 3153-26-2 EINECS: 221-590-8 UN号: 3285 ADR: 6.1 III |
产品技术规格
Appearance | Green to blue-green Crystalline powder |
Infrared spectrum | Conforms |
Assay | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...