锇酸钾

供应商: Thermo Fisher Scientific
Danger

Synonyms: Potassium osmium(VI) oxide dihydrate, Potassium dioxidodioxoosmium dihydrate, Dipotassium osmate dihydrate

197660025 197665000
ACRO197660025EA 0 CNY
ACRO197660025 ACRO197665000
锇酸钾
锇酸钾
分子式: K₂O₄Os·2H₂O
分子量: 368.45 g/mol
MDL编号: MFCD00149919
CAS号: 10022-66-9
UN号: 3085
ADR: 5.1 II

现在订购

产品技术规格

Appearance Violet Crystalline powder
Assay 51.0 to 52.0 % (Os)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR