Torque wrench fittings for HDPE closures, Nalgene®
供应商: Thermo Fisher Scientific
2195-1153 2195-1020 2195-1028 2195-1043 2195-1038 2195-1053 2195-1183
NALG2195-1153EA 0 CNY
NALG2195-1153 NALG2195-1020 NALG2195-1028 NALG2195-1043 NALG2195-1038 NALG2195-1053 NALG2195-1183
Torque wrench fittings for HDPE closures, Nalgene®
扳手
Epoxy, specifically designed for Nalgene HDPE closures.
Proper torquing of Nalgene closures is critical to leakproof sealing.