1-溴-2-氯苯 98.5%
供应商: Thermo Fisher Scientific
|
Warning
|
Synonyms:
2-Bromochlorobenzene
106820250
106821000
ACRO106820250EA
0
CNY
ACRO106820250
ACRO106821000
1-溴-2-氯苯 98.5%
1-溴-2-氯苯
分子式:
C₆H₄BrCl 分子量: 191.45 g/mol 沸点: 204 °C (760 mmHg) 溶点: -12.1 密度: 1.649 g/cm³ 闪点: 70 °C 存储条件: Ambient |
MDL编号:
MFCD00000532 CAS号: 694-80-4 EINECS: 211-775-1 |
产品技术规格
Appearance | Clear colorless to yellow to light orange Liquid (May darken on storage) |
Infrared spectrum | Conforms |
Refractive index | 1.5805 to 1.5825 (20°C, 589 nm) |
GC | ≥98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...