5-溴水杨酸 98%
供应商: Thermo Fisher Scientific
|
Warning
|
Synonyms:
5-溴-2-羟基苯甲酸
162140500
162142500
ACRO162140500EA
0
CNY
ACRO162140500
ACRO162142500
5-溴水杨酸 98%
5-溴水杨酸
分子式:
C₇H₅BrO₃ 分子量: 217.02 g/mol 沸点: 334 溶点: 162…166 °C |
MDL编号:
MFCD00002455 CAS号: 89-55-4 EINECS: 201-917-0 |
产品技术规格
Appearance | White to light beige Powder |
Infrared spectrum | Conforms |
Melting point | 164 to 172 °C |
Titration with NaOH | 97.5 to 102.5 % |
HPLC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...