4-溴苯甲酸 97%

供应商: Thermo Fisher Scientific
Warning

Synonyms: p-Bromobenzoic Acid

148700250 148701000
ACRO148700250EA 0 CNY
ACRO148700250 ACRO148701000
4-溴苯甲酸 97%
4-溴苯甲酸
分子式: C₇H₅BrO₂
分子量: 201.02 g/mol
沸点: 300
溶点: 251…256 °C
存储条件: Ambient
MDL编号: MFCD00002529
CAS号: 586-76-5
EINECS: 209-581-7
Merck Index: 13,01392

现在订购

产品技术规格

Appearance White to yellow to beige Powder
Infrared spectrum Conforms
Melting point 251 to 257 °C
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR