5-溴-2-甲氧基苯甲醛 99%

供应商: Thermo Fisher Scientific
Warning

Synonyms: 5-溴-邻茴香醛

167520250 167520050
ACRO167520250EA 0 CNY
ACRO167520250 ACRO167520050
5-溴-2-甲氧基苯甲醛 99%
5-溴-2-甲氧基苯甲醛
分子式: C₈H₇BrO₂
分子量: 215.05 g/mol
沸点: 269
溶点: 115…118 °C
存储条件: Ambient
MDL编号: MFCD00003347
CAS号: 25016-01-7
EINECS: 246-564-3

现在订购

产品技术规格

Appearance Off-white to yellow to beige Powder or crystals
Infrared spectrum Conforms
Melting point 115 to 119 °C
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR