Petri dishes
供应商: BRAND
452015 452010
BRND452015EA 0 CNY
BRND452015 BRND452010
Petri dishes
培养皿 培养皿
PS, transparent. With or without vent. Dishes and lids are assembled robotically to reduce the risk of contamination, in particular bacterial count.