叔丁胺 99%

供应商: Thermo Fisher Scientific
Danger

Synonyms: 2-氨基-2-甲基丙烷

107820010 107820050
ACRO107820010EA 0 CNY
ACRO107820010 ACRO107820050
叔丁胺 99%
叔丁胺
分子式: C₄H₁₁N
沸点: 46 °C (760 mmHg)
溶点: –67 °C
密度: 0,69 g/cm³
闪点: –38 °C
存储条件: Ambient
MDL编号: MFCD00008050
CAS号: 75-64-9
EINECS: 200-888-1
UN号: 2734
ADR: 8,I
Merck Index: 13,01544

现在订购

产品技术规格

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.3770 to 1.3790 (20°C, 589 nm)
Water ≤0.3 %
GC ≥98.5 %
Color scale ≤10 APHA
SO4 and SO3 (as SO4) ≤0.005 %
Iron (Fe) ≤5 ppm
Heavy metals (as Pb) ≤5 ppm
Titrable acid ≤0.002 meq/g
Insoluble matter ≤0.005 %
Starch Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR