炔丙基胺 99%
供应商: Thermo Fisher Scientific
|
Danger
|
Synonyms:
3-氨基-1-丙炔, 2-丙炔胺
131460050
131460250
ACRO131460050EA
0
CNY
ACRO131460050
ACRO131460250
炔丙基胺 99%
炔丙基胺
分子式:
C₃H₅N 分子量: 55.08 g/mol 沸点: 84 °C (760 mmHg) 密度: 0.86 g/cm³ 闪点: 3 °C 存储条件: Refrigerator |
MDL编号:
MFCD00008198 CAS号: 2450-71-7 EINECS: 219-513-8 UN号: 3286 ADR: 3 II |
产品技术规格
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4460 to 1.4500 (20°C, 589 nm) |
GC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...