1-丁硫醇 99+%

供应商: Thermo Fisher Scientific
Danger

Synonyms: C4 硫醇, 丁基硫醇

220620500 220620050
ACRO220620500EA 0 CNY
ACRO220620500 ACRO220620050
1-丁硫醇 99+%
1-丁硫醇
分子式: C₄H₁₀S
沸点: 98 °C (760 mmHg)
溶点: –116 °C
密度: 0,84 g/cm³
存储条件: Ambient
MDL编号: MFCD00004905
CAS号: 109-79-5
EINECS: 203-705-3
UN号: 2347
ADR: 3,II
Merck Index: 12,01611

现在订购

产品技术规格

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4420 to 1.4440 (20°C, 589 nm)
GC ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR