1-丁硫醇 98%

供应商: Thermo Fisher Scientific
Danger

Synonyms: C4 硫醇, 丁基硫醇

150800500 150802500
ACRO150800500EA 0 CNY
ACRO150800500 ACRO150802500
1-丁硫醇 98%
1-丁硫醇
分子式: C₄H₁₀S
沸点: 98 °C (760 mmHg)
溶点: –116 °C
密度: 0,84 g/cm³
存储条件: Ambient
MDL编号: MFCD00004905
CAS号: 109-79-5
EINECS: 203-705-3
UN号: 2347
ADR: 3,II
Merck Index: 12,01611

现在订购

产品技术规格

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4420 to 1.4440 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR