Compact burettes, SILBERBRAND
供应商: BRAND
10002 10004
BRND10002EA 0 CNY
BRND10002 BRND10004
Compact burettes, SILBERBRAND
滴定管
Soda-lime glass AR-GLAS®.
  • Calibrated to deliver (TD, Ex)
  • Black graduations
  • Discharge tube with outer diameter 8 mm
Error limits according to class B.