3-戊醇 98%

供应商: Thermo Fisher Scientific
Warning

Synonyms: 二乙基甲醇, sec-inact.-Pentyl alcohol

129990250 129991000
ACRO129990250EA 0 CNY
ACRO129990250 ACRO129991000
3-戊醇 98%
3-戊醇
分子式: C₅H₁₂O
沸点: 114…116 °C (760 mmHg)
溶点: –75 °C
密度: 0,815 g/cm³
闪点: 30 °C
存储条件: Ambient
MDL编号: MFCD00004574
CAS号: 584-02-1
EINECS: 209-526-7
UN号: 1105
ADR: 3,III
Merck Index: 13,07197
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2ec7192926b50615dfac930445c63174c6221a44&vid=a2f1fa3f1ffa1ecbfb6a5dd1d0f29da27d1d8737&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4090 to 1.4110 (20°C, 589 nm)
GC ≥97.5 %
Iron (Fe) ≤5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR