1-戊醇 ≥98%, GPR RECTAPUR®

供应商: VWR Chemicals
Warning

Synonyms: 戊醇, 1-戊醇, C5醇, 正戊醇

VWRC20800.294EA 491 CNY
VWRC20800.294
1-戊醇 ≥98%, GPR RECTAPUR®
1-戊醇
分子式: H₃C(CH₂)₃CH₂OH
分子量: 88.15 g/mol
沸点: 137.9 °C (1013 hPa)
溶点: –79 °C
密度: 0.81 g/cm³ (20 °C)
闪点: 48
存储条件: Ambient
MDL编号: MFCD00002977
CAS号: 71-41-0
EINECS: 200-752-1
UN号: 1105
ADR: 3 III
Merck Index: 13,07195
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=92dac1192eb8e068fc1804fdcf488129a2385918&vid=d4c7f2c5672a97c6dfaddc50807d768068096252&prid=web&clid=SAPDAM

现在订购

产品技术规格

Assay Min. 98 %
Identification (IR) Passes test
Density (20/4) 0.813 --> 0.817
Refractive index (20°C) 1.409 --> 1.412
Water (KF) Max. 0.30 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR