1-辛醇 99%, pure

供应商: Thermo Fisher Scientific
Warning

Synonyms: C8 醇, 正辛醇, 辛醇

150630010 150630025
ACRO150630010EA 0 CNY
ACRO150630010 ACRO150630025
1-辛醇 99%, pure
1-辛醇
分子式: H₃C(CH₂)₆CH₂OH
沸点: 195 °C (760 mmHg)
溶点: –16 °C
密度: 0,824 g/cm³
闪点: 81 °C
存储条件: Ambient
MDL编号: MFCD00002988
CAS号: 111-87-5
EINECS: 203-917-6
Merck Index: 13,06784
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7db4a6be3aedee4af43781ec6deb31631bc376c1&vid=80cb13f618a2d3181daf8310324993f1f7c69fb2&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4280 to 1.4310 (20°C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR