正庚醇 98%

供应商: Thermo Fisher Scientific
Warning

Synonyms: 庚醇

120360010 120362500
ACRO120360010EA 0 CNY
ACRO120360010 ACRO120362500
正庚醇 98%
正庚醇
分子式: H₃C(CH₂)₅CH₂OH
沸点: 176 °C (760 mmHg)
溶点: –35 °C
密度: 0,8219 g/cm³
闪点: 73 °C
存储条件: Ambient
MDL编号: MFCD00002986
CAS号: 111-70-6
EINECS: 203-897-9
Merck Index: 13,04681

现在订购

产品技术规格

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4230 to 1.4250 (20°C, 589 nm)
Water ≤0.5 %
GC ≥97.5 %
Color scale ≤30 APHA
Residue after evaporation ≤0.01 %
Bromine number ≤0.1 g Br/100 g
Acid number ≤0.5 mg KOH/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR