1-十七烷醇 97%

供应商: Thermo Fisher Scientific

Synonyms: 十七醇

221230050 221230250
ACRO221230050EA 0 CNY
ACRO221230050 ACRO221230250
1-十七烷醇 97%
1-十七烷醇
分子式: H₃C(CH₂)₁₅CH₂OH
分子量: 256.47 g/mol
沸点: 308.5 °C (760 mmHg)
溶点: 51…56 °C
闪点: 154 °C
存储条件: Ambient
MDL编号: MFCD00002822
CAS号: 1454-85-9
EINECS: 215-932-5

现在订购

产品技术规格

Appearance White to off-white Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 51 to 56 °C
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR