正丙胺 99+%, extra pure
供应商: Thermo Fisher Scientific
|
Danger
|
Synonyms:
1-氨基丙烷
220610010
220610500
220615000
ACRO220610010EA
0
CNY
ACRO220610010
ACRO220610500
ACRO220615000
正丙胺 99+%, extra pure
正丙胺
分子式:
H₃CCH₂CH₂NH₂ 分子量: 59.11 g/mol 沸点: 48 °C (760 mmHg) 溶点: –83 °C 密度: 0.719 g/cm³ 闪点: –30 °C 存储条件: Refrigerator |
MDL编号:
MFCD00008205 CAS号: 107-10-8 EINECS: 203-462-3 UN号: 1277 ADR: 3 II Merck Index: 12,08028 |
产品技术规格
Appearance | Clear liquid |
Infrared spectrum | Conforms |
Refractive index | 1.3870 to 1.3890 (20°C, 589 nm) |
Water | ≤0.2 % (K.F.) |
GC | ≥99.0 % |
Color scale | ≤20 APHA |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...