十八烷基胺 90% tech.

供应商: Thermo Fisher Scientific
Danger

Synonyms: 1-氨基十八烷, 硬脂胺

129320010 129325000
ACRO129320010EA 0 CNY
ACRO129320010 ACRO129325000
十八烷基胺 90% tech.
十八烷基胺
分子式: H₃C(CH₂)₁₆CH₂NH₂
沸点: 232 °C (32 mmHg)
溶点: 50…60 °C
密度: 0,8618 g/cm³ (20 °C)
闪点: 148 °C
存储条件: Ambient
MDL编号: MFCD00008159
CAS号: 124-30-1
EINECS: 204-695-3
UN号: 3077
ADR: 9,III

现在订购

产品技术规格

Appearance White Pastilles
Infrared spectrum Conforms
Melting point 50 to 60 °C
Water ≤0.5 %
GC ≥88 %
Titration with HCl ≥2 % (sec.+ tert.amine MW 521)
Iodine value ≤4 g I/100 g
Amine number 204 to 211 mg KOH/g (Total)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR