十二胺 98%
供应商: Thermo Fisher Scientific
|
Danger
|
Synonyms:
1-氨基十二烷, 月桂胺
117660010
117661000
117665000
ACRO117660010EA
0
CNY
ACRO117660010
ACRO117661000
ACRO117665000
十二胺 98%
十二胺
分子式:
H₃C(CH₂)₁₀CH₂NH₂ 沸点: 247 °C (1013 hPa) 溶点: 28…32 °C 密度: 0,8 g/cm³ (20 °C) 闪点: >110 °C 存储条件: Ambient |
MDL编号:
MFCD00008154 CAS号: 124-22-1 EINECS: 204-690-6 UN号: 3259 ADR: 8,II |
产品技术规格
Appearance | White Low melting crystalline solid |
Infrared spectrum | Conforms |
Melting point | 27 to 31 °C |
GC | ≥97.5 % (Alkyl chain distribution C12) |
Titration with HClO4 | ≥97.5 % |
Amine number, total | 297 to 304 mg KOH/g |
Residue after evaporation | ≤0.005 % |
Titrable acid | ≤0.002 meq/g |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...