Tributylamine 99%
供应商: Thermo Fisher Scientific
|
Danger
|
139320010
139321000
ACRO139320010EA
0
CNY
ACRO139320010
ACRO139321000
Tributylamine 99%
Tributylamine
分子式:
C₁₂H₂₇N 分子量: 185.35 g/mol 沸点: 215 °C (760 mmHg) 溶点: –70 °C 密度: 0.778 g/cm³ 闪点: 75 °C 存储条件: Ambient |
MDL编号:
MFCD00009431 CAS号: 102-82-9 EINECS: 203-058-7 UN号: 2542 ADR: 6.1 II Merck Index: 13,09691 |
产品技术规格
Appearance | Clear liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4273 to 1.4293 (20°C, 589 nm) |
Water | ≤0.2 % |
GC | ≥98.5 % |
Color scale | ≤20 APHA |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...