庚酸 98%
供应商: Thermo Fisher Scientific
|
Danger
|
Synonyms:
葡萄花酸, 正庚酸
164170010
164172500
ACRO164170010EA
0
CNY
ACRO164170010
ACRO164172500
庚酸 98%
庚酸
分子式:
C₇H₁₄O₂ 沸点: 223…223,4 °C (760 mmHg) 密度: 0,91 g/cm³ 闪点: >112 °C 存储条件: Ambient |
MDL编号:
MFCD00004426 CAS号: 111-14-8 EINECS: 203-838-7 UN号: 3265 ADR: 8,III Merck Index: 13,04680 |
产品技术规格
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4210 to 1.4250 (20°C, 589 nm) |
Titration with NaOH | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...