乙酸丙酯 98,0 GC_ASSAY_METHOD
供应商: TCI
|
Danger
|
Synonyms:
Acetic acid propyl ester
A0044-25ML
A0044-500ML
TCIAA0044-25MLEA
0
CNY
TCIAA0044-25ML
TCIAA0044-500ML
乙酸丙酯 98,0 GC_ASSAY_METHOD
乙酸丙酯
分子式:
C₅H₁₀O₂ 分子量: 102.13 g/mol 沸点: 101°C 溶点: -95 闪点: 13°C 存储条件: Ambient |
MDL编号:
MFCD00009372 CAS号: 109-60-4 EINECS: 203-686-1 UN号: 1276 ADR: 3 II Merck Index: 14,07841 |
|
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...