3,5-庚烷二酮 99%
供应商: Thermo Fisher Scientific
|
Warning
|
411260050
411260250
ACRO411260050EA
0
CNY
ACRO411260050
ACRO411260250
3,5-庚烷二酮 99%
3,5-庚烷二酮
分子式:
C₇H₁₂O₂ 分子量: 128.17 g/mol 沸点: 172…173 °C (711 mmHg) 密度: 0.945 g/cm³ 闪点: 57 °C 存储条件: Ambient |
MDL编号:
MFCD00015186 CAS号: 7424-54-6 EINECS: 231-054-5 UN号: 1224 ADR: 3 III |
产品技术规格
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4540 to 1.4570 (20°C, 589 nm) |
GC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...