2-壬酮 99%

供应商: Thermo Fisher Scientific
Warning

Synonyms: 甲基庚基酮

148780050 148781000
ACRO148780050EA 0 CNY
ACRO148780050 ACRO148781000
2-壬酮 99%
2-壬酮
分子式: C₉H₁₈O
分子量: 142.24 g/mol
沸点: 188…190 °C (760 mmHg)
溶点: –21 °C
密度: 0.826 g/cm³
闪点: 68 °C
存储条件: Ambient
MDL编号: MFCD00009553
CAS号: 821-55-6
EINECS: 212-480-0

现在订购

产品技术规格

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4200 to 1.4220 (20°C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR