三丁基乙烯基锡 97%
供应商: Thermo Fisher Scientific
|
Danger
|
Synonyms:
乙烯基三丁基锡, 三正丁基(乙烯基)锡, 三丁基乙烯基锡
350000010
350000050
ACRO350000010EA
0
CNY
ACRO350000010
ACRO350000050
三丁基乙烯基锡 97%
三丁基乙烯基锡
分子式:
C₁₄H₃₀Sn 沸点: 104…106 °C (3,5 mmHg) 密度: 1,08 g/cm³ 闪点: >110 °C 存储条件: Ambient |
MDL编号:
MFCD00009421 CAS号: 7486-35-3 EINECS: 231-291-4 UN号: 2788 ADR: 6.1,III |
产品技术规格
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4770 to 1.4790 (20°C, 589 nm) |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...