4-Nitrophenyl acetate 97%
供应商: Thermo Fisher Scientific
|
Danger
|
128750250
128751000
128750050
ACRO128750250EA
0
CNY
ACRO128750250
ACRO128751000
ACRO128750050
4-Nitrophenyl acetate 97%
4-Nitrophenyl acetate
分子式:
C₈H₇NO₄ 溶点: 76…79 °C 存储条件: Freezer |
MDL编号:
MFCD00007326 CAS号: 830-03-5 EINECS: 212-593-5 |
产品技术规格
Appearance | Yellow to beige Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 76 to 79 °C |
GC | ≥96.0 % |
Specific optical rotation | +23° to +29° (20 °C, 589 nm) (c=1 in Chloroform) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...