2-甲基乙酰乙酸乙酯 95%
供应商: Thermo Fisher Scientific
Synonyms:
Ethyl 2-methyl-3-oxobutanoate
326110250
326111000
ACRO326110250EA
0
CNY
ACRO326110250
ACRO326111000
2-甲基乙酰乙酸乙酯 95%
2-甲基乙酰乙酸乙酯
分子式:
C₇H₁₂O₃ 分子量: 144.17 g/mol 沸点: 187 溶点: –45 °C 密度: 1.01 g/cm³ 闪点: 62 °C |
MDL编号:
MFCD00009164 CAS号: 609-14-3 EINECS: 210-179-9 |
产品技术规格
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4170 to 1.4230 (20°C, 589 nm) |
Water | ≤1 % (K.F.) |
GC | ≥94 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...