2,2,2-Trifluorothanesulphonyl chloride 99%

供应商: Thermo Fisher Scientific
Danger

Synonyms: 2,2,2-三氟乙烷磺酰氯

305350010 305350025
ACRO305350010EA 0 CNY
ACRO305350010 ACRO305350025
2,2,2-Trifluorothanesulphonyl chloride 99%
2,2,2-三氟乙基磺酰氯
分子式: C₂H₂ClF₃O₂S
分子量: 182.55 g/mol
沸点: 64…67 °C (45 mmHg)
密度: 1.65 g/cm³
闪点: 72 °C
存储条件: Refrigerator
MDL编号: MFCD00007458
CAS号: 1648-99-3
EINECS: 216-713-7
UN号: 3265
ADR: 8 II

现在订购

产品技术规格

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3870 to 1.3890 (20°C, 589 nm)
Titration Argentometric ≥98.5 % (ex Cl)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR