4-乙基愈创木酚 98%

供应商: Thermo Fisher Scientific
Warning

Synonyms: 5-Ethyl-2-hydroxyanisole, 1-Hydroxy-2-methoxy-4-ethyl Benzene, 4-Ethylguaiacol

351960050 351960250
ACRO351960050EA 0 CNY
ACRO351960050 ACRO351960250
4-乙基愈创木酚 98%
4-乙基愈创木酚
分子式: C₉H₁₂O₂
分子量: 152.19 g/mol
沸点: 234…236 °C
溶点: 15 °C
闪点: 107 °C
MDL编号: MFCD00038714
CAS号: 2785-89-9
EINECS: 220-500-4

现在订购

产品技术规格

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5250 to 1.5300 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR