甘油, Sigma-Aldrich®

供应商: Merck


M-Clarity™ quality level = MQ200

G5516-100ML G5516-500ML
SIALG5516-100MLEA 0 CNY
SIALG5516-100ML SIALG5516-500ML
甘油, Sigma-Aldrich®
甘油
分子式: HOCH₂CH(OH)CH₂OH
沸点: 290 °C (1013 hPa)
密度: 1,26 g/cm³ (20 °C)
闪点: 177 °C
存储条件: Ambient
MDL编号: MFCD00004722
CAS号: 56-81-5
EINECS: 200-289-5
Merck Index: 13,04497
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3668cce85af1464db21fd098526252076b567956&vid=86a15453b54e7106c121d32e4e716385003c4507&prid=web&clid=SAPDAM

现在订购


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR