酪胺 ≥98.0% (by GC, titration analysis)
供应商: TCI
|
Warning
|
Synonyms:
4-(2-氨乙基)苯酚, 4-羟基苯乙胺, 2-(4-羟基苯基)乙胺
A0302-25G
A0302-250G
TCIAA0302-25GEA
0
CNY
TCIAA0302-25G
TCIAA0302-250G
酪胺 ≥98.0% (by GC, titration analysis)
酪胺
分子式:
C₈H₁₁NO 分子量: 137.18 g/mol 沸点: 325 溶点: 164 |
MDL编号:
MFCD00008193 CAS号: 51-67-2 EINECS: 200-115-8 Merck Index: 14,09835 |
|
产品技术规格
Appearance | White to Light yellow to Light orange powder to crystaline |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 160.0 to 165.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...