1-溴十四烷 98%

供应商: Thermo Fisher Scientific

Synonyms: 肉豆蔻基溴, 十四烷基溴

187381000 187385000
ACRO187381000EA 0 CNY
ACRO187381000 ACRO187385000
1-溴十四烷 98%
1-溴十四烷
分子式: BrH₂C(CH₂)₁₂CH₃
沸点: 175…178 °C (20 mmHg)
溶点: 5…6 °C
密度: 0,93 g/cm³
闪点: 149 °C
存储条件: Ambient
MDL编号: MFCD00000228
CAS号: 112-71-0
EINECS: 203-999-3

现在订购

产品技术规格

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4580 to 1.4620 (20°C, 589 nm)
GC ≥97.5 %
Ethanol 95 to 97 % v/v (calculated)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR