1-Bromobutane 99%
供应商: Thermo Fisher Scientific
|
Danger
|
Synonyms:
Butyl bromide
106770010
106772500
ACRO106770010EA
0
CNY
ACRO106770010
ACRO106772500
1-Bromobutane 99%
1-Bromobutane
分子式:
BrH₂C(CH₂)₂CH₃ 分子量: 137.02 g/mol 沸点: 100…104 °C (760 mmHg) 溶点: –112 °C 密度: 1.27 g/cm³ 闪点: 10 °C 存储条件: Ambient |
MDL编号:
MFCD00000260 CAS号: 109-65-9 EINECS: 203-691-9 UN号: 1126 ADR: 3 II Merck Index: 13,01552 |
产品技术规格
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4384 to 1.4404 (20°C, 589 nm) |
GC | ≥98.5 % |
Gravimetry | ≥99.0 % |
Suitability | (for Ni determination) Passes test |
Insoluble matter | ≤0.05 % (in ethanol) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...