硫代硫酸钠 0.1 N (N/10) in aqueous solution, StandARd® volumetric solution, Macron Fine Chemicals™
供应商: Avantor Performance Materials
Synonyms:
Sodiumthiosulphate, Sodium hyposulphite, Antichlor, di-Sodium thiosulphate
MACR4682-60EA
752
CNY
MACR4682-60
MACR4682-01
硫代硫酸钠 0.1 N (N/10) in aqueous solution, StandARd® volumetric solution, Macron Fine Chemicals™
硫代硫酸钠
分子式:
Na₂S₂O₃ 分子量: 158.11 g/mol 沸点: 100 溶点: 48 存储条件: Ambient |
MDL编号:
MFCD00003499 CAS号: 7772-98-7 EINECS: 231-853-9 ADR: 9 III Merck Index: 12,08844 |
产品技术规格
For Laboratory,Research,or Manufacturing Use | — |
Normality | 0.0995 - 0.1005 |