(+)-葑酮 97%
供应商: Thermo Fisher Scientific
Synonyms:
(+)-1,3,3-三甲基-2-降冰片酮, (1S)-1,3,3-三甲基二环[2.2.1]庚-2-酮
366290250
366291000
ACRO366290250EA
0
CNY
ACRO366290250
ACRO366291000
(+)-葑酮 97%
(+)-葑酮
分子式:
C₁₀H₁₆O 分子量: 152.24 g/mol 沸点: 63…65 °C (13 mmHg) 溶点: 5…7 °C 密度: 0.95 g/cm³ (20 °C) 闪点: 62 °C 存储条件: Ambient |
MDL编号:
MFCD00070689 CAS号: 4695-62-9 EINECS: 225-160-0 Merck Index: 13,03995 |
产品技术规格
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4610 to 1.4640 (20°C, 589 nm) |
GC | ≥96.0 % |
Specific optical rotation | +55° to +63° (20°C, 589 nm) (neat) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...