Centrifuge tubes, cylindrical, round bottom
供应商: BRAND
115342 115346 115348 115350 115352 115354 115356
BRND115342EA 0 CNY
BRND115342 BRND115346 BRND115348 BRND115350 BRND115352 BRND115354 BRND115356
Centrifuge tubes, cylindrical, round bottom
PP, translucent.
  • Translucent, rimmed, very good chemical resistance
  • Autoclavable at 121 °C (2 bar), according DIN EN 285
  • Fitting stoppers must be ordered separately