Single-use syringes, 3-piece, HENKE-JECT®
供应商: HENKE SASS WOLF
8300005761 5050.000V0 5200.X00V0 8300006682 8300005458 5300.X00D0
HSWA8300005761EA 0 CNY
HSWA8300005761 HSWA5050.000V0 HSWA5200.X00V0 HSWA8300006682 HSWA8300005458 HSWA5300.X00D0
Single-use syringes, 3-piece, HENKE-JECT®
注射器 医用注射器
Disposable, three-piece syringes with PP barrel and piston, polyisoprene stopper, latex-free.
  • Lubricated synthetic rubber piston allows a smooth draw
  • Available in non sterile version (bulk) upon request
Without needles: Luer lock types are compatible for use with most common syringe pumps. Please ask for further information.
With fixed needles: Needle tube permanently attached in syringe, no dead space, safety stop, suitable for dosage of very small volumes.
Luer lock types are compatible for use with most common syringe pumps. Please ask for further information.
Packaging: Individual sterile blisters.