Chemical resistant overalls, with integrated socks, Tychem® SL
供应商: DUPONT PROTECTIVE APPAREL
SL122BWHXL00 SL122BWH3X00
DUPOSL122BWHXL00EA 0 CNY
DUPOSL122BWHXL00 DUPOSL122BWH3X00
Chemical resistant overalls, with integrated socks, Tychem® SL
Disposable Garments
These overalls provide effective protection against a range of chemical environments. Tychem® SL, utilising Saranex® 23-P film laminated to Tyvek® brand protective material, is a lightweight, comfortable garment specifically designed for easy wear. It provides at least 30 min of protection against >124 chemical challenges.
  • Tightly sewn seam is covered with garment fabric to reinforce seam and reduce potential for particle penetration
  • Attached hood with elastic around face opening
  • Storm flap covers zip which can be sealed by the wearer with adhesive strip to prevent intrusion at zip
  • Elastic opening for tighter fit at wrist
  • Integrated socks composed of garment material
Typical applications include chemical mixing, hazardous remediation, emergency medical response, paint spraying, and nuclear environments.
Colour: White