过氧化氢 30% stabilised Ph. Eur., USP, BP
供应商: VWR Chemicals
|
Danger
|
Stabilization:
Organic stabilisers ≤500 ppm
VWRC23622.467EA
0
CNY
VWRC23622.467
VWRC23622.367
VWRC23622.298
VWRC23622.330
VWRC23622.260
过氧化氢 30% stabilised Ph. Eur., USP, BP
过氧化氢
Caution: Not for use as Active Pharmaceutical Ingredient (API).
Not for use as disinfectant. Not registered as biocide.
分子式:
H₂O₂ 分子量: 34,01 g/mol 沸点: ~107 °C (1013 hPa) 溶点: <0 °C 密度: 1,11 g/cm³ (20 °C) 存储条件: Ambient |
MDL编号:
MFCD00011333 CAS号: 7722-84-1 EINECS: 231-765-0 UN号: 2014 ADR: 5.1 (8),II |
产品技术规格
Appearance | Clear colourless liquid |
Residual solvents | Passes test |
As (Arsenic) | ≤ 2 ppm |
Cd (Cadmium) | ≤ 1 ppm |
Co (Cobalt) | ≤ 1 ppm |
Hg (Mercury) | ≤ 2 ppm |
Ni (Nickel) | ≤ 1 ppm |
Pb (Lead) | ≤ 2 ppm |
V (Vanadium) | ≤ 1 ppm |
Conforms to Ph.Eur./USP/BP | Passes test |
Ph.Eur./BP specifications: | |
Assay [Ph.Eur./BP] | 29.0 - 31.0 % |
Identification A | Passes test Ph.Eur./BP |
Identification B | Passes test Ph.Eur./BP |
Identification C | Passes test Ph.Eur./BP |
Acidity | Passes test Ph.Eur./BP |
Organic stabilizers [Ph.Eur./BP] | ≤ 500 ppm |
Non-volatile residue [Ph.Eur./BP] | ≤ 2 g/l |
USP specifications: | |
Assay [USP] | 29.0 - 32.0 % |
Identification A | Passes test USP |
Cl (Chloride) [USP] | ≤ 50 ppm |
Limit of nonvolatile residue [USP] | ≤ 30 mg |
Acidity | Passes test USP |
Limit of preservative [USP] | ≤ 0.05 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...