丙酮-D6 (99.9% D)

供应商: CAMBRIDGE ISOTOPE LABS
Danger

Synonyms: 氘代丙酮

DLM-9-10 DLM-9
CAMBDLM-9-10EA 0 CNY
CAMBDLM-9-10 CAMBDLM-9
丙酮-D6 (99.9% D)
丙酮-D6
分子式: D₃CC(O)CD₃
分子量: 64.03 g/mol
沸点: 55.5
溶点: –95.4 °C
密度: 0.79 g/cm³ (20 °C)
闪点: –19 °C (closed cup)
存储条件: Ambient
MDL编号: MFCD00044635
CAS号: 666-52-4
EINECS: 211-563-9
UN号: 1090
ADR: 3 II

现在订购


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR