Cleanroom gloves, latex, Kimtech™ G5
供应商: KIMBERLY CLARK
HC220 HC330 HC440 HC3411
KIMBHC220EA 0 CNY
KIMBHC220 KIMBHC330 KIMBHC440 KIMBHC3411
Cleanroom gloves, latex, Kimtech™ G5
手套
Ambidextrous gloves with beaded cuffs for easy donning and better seal on garment. Ideal for handling delicate equipment in ISO Class 5 or higher cleanrooms within microelectronics, semiconductor, optics and pharmaceutical industries.
  • Non sterile, powder-free
  • Fifty micrograms per gram (50 µg/g) or less of total water extractable protein
  • Manufactured from natural rubber latex for tactile sensitivity and comfort
  • Textured palm and fingers for better grip
  • Silicone-free material
  • Virus resistance according to ISO 16604
Suitable for use in Class 100, ISO 5, M3.5 or higher cleanrooms.
EN 374; Cat. III; AQL 1.5
Certifications: PPE Cat III (EU) 2016/425, EN 374-1 Type C (K), EN 374-5 (VIRUS), AQL 1,5
Packaging: 100 gloves, double bagged in sealed cleanroom polyethylene bags.