Cleanroom overalls, Kimtech™ A5
供应商: KIMBERLY CLARK
88801 88802 88803 88804 88805
KIMB88801EA 0 CNY
KIMB88801 KIMB88802 KIMB88803 KIMB88804 KIMB88805
Cleanroom overalls, Kimtech™ A5
Disposable Garments
Kimtech™ A5 sterile cleanroom apparel with CLEAN-DON™ technology is made of SMS (Spunbond Melt-blown Spunbond) material. It features built-in snaps that gather up legs and arms to aid aseptic gowning. Garments have a thumb loop that prevents the sleeve from rolling back and exposing arm or wrist when in use.
  • Cloth-like comfort
  • Innovative inside-out fold pattern - reduces the risk of touching and contaminating the outside of the apparel when opening or donning
  • Highly visible blue line along the inside of the garment signals the proper place to grasp while gowning
  • Vacuum packaging reduces space, so more products fit into gowning area storage
Suitable for use in Class 100, ISO 5, 3.5 environments.
Colour: White
Certifications: PPE Cat III (EU) 2016/425, Type 5/6
Packaging: Vacuum packed and double-bagged for added sterility assurance.