Detergent, Alconox® powder
供应商: Alconox
1104 1150
ALCO1104EA 0 CNY
ALCO1104 ALCO1150
Detergent, Alconox® powder
清洗剂 洗涤剂和表面清洁剂
Concentrated detergent is ideal for removing soil, grit, buffing compounds, and other organic contaminants from glass, metal, stainless steel, porcelain, ceramic, plastic, rubber, and fiberglass. Non corrosive, high-foaming formula can be used on soft metals. Consists of homogeneous blend of sodium linear alkylaryl sulfonate, alcohol sulfate, phosphates, and carbonates.
  • Corrosion inhbited
  • Free-rinsing, odorless, anionic, and mild (pH 9,5)
  • Typical dosage: Dilute 1:100