VWR® Sharps Container Systems, Yellow
供应商: VWR BRAND PRIVATE LABEL USA
19001-041 19001-039
VWRU19001-041EA 115 CNY
VWRU19001-041 VWRU19001-039
VWR® Sharps Container Systems, Yellow
箱柜 Sharps Containers
Sharps container systems provide solutions for the safe and effective containment and disposal of contaminated needles, syringes and other sharps.
  • Stackable
  • Puncture-resistant
  • Autoclavable
  • Transparent, locking lids
Containers are stackable, puncture-resistant, and autoclave- and incinerator-safe. They feature transparent, locking lids and needle removal ports. The tortuous path systems restrict hand entry.