VWR® Testing Sieves, All Stainless Steel
供应商: VWR BRAND PRIVATE LABEL USA
57334-480 57334-452 57334-456 57334-460 57334-478 57334-486 57334-470 57334-444 57334-454 57334-458 57334-464 57334-474 57334-476 57334-482 57334-484 57334-488 57334-494 57334-448 57334-450 57334-446 57334-434 57334-438 57334-436 57334-440 57334-442 57334-428 57334-432 57334-562 57334-566 57334-574 57334-578 57334-570 57334-586 57334-590 57334-592 57334-582 57334-596 57334-598 57334-466 57334-472
VWRU57334-480EA 4707.6 CNY
VWRU57334-480 VWRU57334-452 VWRU57334-456 VWRU57334-460 VWRU57334-478 VWRU57334-486 VWRU57334-470 VWRU57334-444 VWRU57334-454 VWRU57334-458 VWRU57334-464 VWRU57334-474 VWRU57334-476 VWRU57334-482 VWRU57334-484 VWRU57334-488 VWRU57334-494 VWRU57334-448 VWRU57334-450 VWRU57334-446 VWRU57334-434 VWRU57334-438 VWRU57334-436 VWRU57334-440 VWRU57334-442 VWRU57334-428 VWRU57334-432 VWRU57334-562 VWRU57334-566 VWRU57334-574 VWRU57334-578 VWRU57334-570 VWRU57334-586 VWRU57334-590 VWRU57334-592 VWRU57334-582 VWRU57334-596 VWRU57334-598 VWRU57334-466 VWRU57334-472
VWR® Testing Sieves, All Stainless Steel
筛子 试验筛
Brass wire cloth and frame for higher corrosion resistance than sieves containing brass wire cloth or frames. High durability and wear resistance. Less likelihood of metallic sample contamination. Full coarse and fine mesh range down to 20 µm (0.0008"). Sturdy construction with fillet solder bead between the frame and wire cloth to prevent particles from being trapped. Diameter: 20,3 cm (8"). Choose full height with 5,08cm (2") depth to wire, or half height with 2,54 cm (1") depth to wire.
Inspected with an optical measurement system traceable to the National Institute of Standards and Technology. Each sieve bears a traceable serial number and comes with certificate of compliance to applicable ASTM E-11 and ISO specifications.
CENTERLINE® measurement (47730-590) is a NIST traceable service that checks calibration and gives statistical measurement information on each sieve.